Search
Search

RMDN2| Gene Symbol: RMDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144713.4 | NP_653314.3 | 2 | 759 | ||
| XM_011532613.2 | XP_011530915.1 | 2 | 740 | |||
| XM_011532614.2 | XP_011530916.1 | 4 | 871 | |||
| XM_011532615.2 | XP_011530917.2 | 4 | 871 | |||
| XM_017003474.1 | XP_016858963.1 | 4 | 871 | |||
| XM_017003475.1 | XP_016858964.1 | 4 | 871 | |||
| GenBank mRNA | AK057516.1 | - | 2 | 741 | ||