Search
Search

FANCD2OS| Gene Symbol: FANCD2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164839.1 | NP_001158311.1 | 2 | 475 | ||
| NM_173472.1 | NP_775743.1 | 2 | 475 | |||
| XM_011533330.2 | XP_011531632.1 | 3 | 943 | |||
| XM_011533331.2 | XP_011531633.1 | 2 | 618 | |||
| GenBank mRNA | AF230334.1 | - | 2 | 467 | ||
| AY248899.1 | - | 2 | 325 | |||
| BC028122.1 | - | 2 | 475 | 5168387 | ||