Search
Search

EOGT| Gene Symbol: EOGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278689.1 | NP_001265618.1 | 5 | 977 | ||
| NM_173654.2 | NP_775925.1 | 5 | 977 | |||
| NR_103826.1 | 5 | 977 | ||||
| XM_005264743.2 | XP_005264800.1 | 5 | 1191 | |||
| XM_011533599.1 | XP_011531901.1 | 5 | 1191 | |||
| XM_011533600.1 | XP_011531902.1 | 5 | 1044 | |||
| XM_011533602.1 | XP_011531904.1 | 5 | 956 | |||
| XM_011533603.1 | XP_011531905.1 | 5 | 1191 | |||
| XM_017006204.1 | XP_016861693.1 | 5 | 1044 | |||
| XM_017006205.1 | XP_016861694.1 | 5 | 956 | |||
| XM_017006206.1 | XP_016861695.1 | 5 | 852 | |||
| XM_017006208.1 | XP_016861697.1 | 5 | 1044 | |||
| GenBank mRNA | AJ868234.1 | - | 2 | 301 | ||
| AK126187.1 | - | 4 | 448 | |||
| AK290356.1 | - | 1 | 104 | |||
| AK304102.1 | - | 5 | 733 | |||
| BC060887.1 | - | 5 | 706 | 30347170 | ||
| BX640821.1 | - | 1 | 98 | |||
| BX648818.1 | - | 5 | 752 | |||
| KC347596.1 | - | 2 | 301 | |||