Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198476.3 | NP_940878.3 | 6 | 2486 | ||
XM_005258776.4 | XP_005258833.1 | 6 | 3056 | |||
XM_005258778.4 | XP_005258835.1 | 6 | 2475 | |||
XM_011526774.2 | XP_011525076.1 | 8 | 2533 | |||
XM_011526776.2 | XP_011525078.1 | 8 | 2461 | |||
XM_011526777.2 | XP_011525079.1 | 7 | 2416 | |||
XM_011526778.2 | XP_011525080.1 | 7 | 2344 | |||
XM_011526779.2 | XP_011525081.1 | 7 | 1929 | |||
XM_011526783.2 | XP_011525085.1 | 6 | 2143 | |||
XM_011526784.2 | XP_011525086.1 | 9 | 1856 | |||
XM_011526785.2 | XP_011525087.1 | 8 | 1942 | |||
XM_017026620.1 | XP_016882109.1 | 9 | 2207 | |||
XM_017026621.1 | XP_016882110.1 | 8 | 2162 | |||
XM_017026622.1 | XP_016882111.1 | 6 | 1725 | |||
XM_017026623.1 | XP_016882112.1 | 5 | 2026 | |||
XM_017026624.1 | XP_016882113.1 | 9 | 1485 | |||
XM_017026626.1 | XP_016882115.1 | 7 | 2389 | |||
XM_017026627.1 | XP_016882116.1 | 5 | 2942 | |||
GenBank mRNA | AK123126.1 | - | 9 | 1475 | ||
BC090894.1 | - | 6 | 2252 | 30377779 |