Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270934.1 | NP_001257863.1 | 6 | 1505 | ||
NM_001270941.1 | NP_001257870.1 | 6 | 1505 | |||
NM_001282282.1 | NP_001269211.1 | 5 | 1228 | |||
NM_014790.4 | NP_055605.2 | 6 | 1505 | |||
XM_017010097.1 | XP_016865586.1 | 7 | 1499 | |||
XM_017010098.1 | XP_016865587.1 | 6 | 1505 | |||
GenBank mRNA | AB011127.1 | - | 6 | 1199 | ||
AF273057.1 | - | 6 | 1346 | |||
AK291280.1 | - | 6 | 1227 | |||
AK299724.1 | - | 5 | 1154 | |||
BC003189.1 | - | 6 | 1151 | |||
BC017354.2 | - | 6 | 1270 | 5020741 | ||
EF512550.1 | - | 5 | 964 |