Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RMDN1 SIRNA Assays ›
          Gene Symbol:
          RMDN1
          Gene Name:
          regulator of microtubule dynamics 1
          Gene Aliases:
          CGI-90, FAM82B, RMD-1, RMD1
          Chromosome Location:
          Chr.8: 86467116 - 86514348 on Build GRCh38
          UniGene ID:
          Hs.145386
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 126875

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RMDN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286707.1 NP_001273636.1 5, 6 705
          NM_001286719.1 NP_001273648.1 5, 6 705
          NM_001317807.1 NP_001304736.1 6, 7 613
          NM_016033.2 NP_057117.2 6, 7 795
          XM_005250944.3 XP_005251001.1 6, 7 701
          XM_005250947.3 XP_005251004.1 5, 6 611
          XM_005250948.3 XP_005251005.1 6, 7 603
          XM_006716573.3 XP_006716636.1 6, 7 790
          XM_011517090.2 XP_011515392.1 6, 7 775
          XM_011517091.2 XP_011515393.1 6, 7 775
          XM_011517092.2 XP_011515394.1 5, 6 685
          XM_011517093.2 XP_011515395.1 6, 7 775
          XM_011517094.2 XP_011515396.1 6, 7 775
          XM_011517095.2 XP_011515397.1 6, 7 775
          XM_011517096.2 XP_011515398.1 6, 7 775
          XM_011517097.2 XP_011515399.1 5, 6 685
          XM_011517098.2 XP_011515400.1 5, 6 685
          XM_017013518.1 XP_016869007.1 6, 7 701
          XM_017013519.1 XP_016869008.1 6, 7 775
          XM_017013520.1 XP_016869009.1 6, 7 701
          XM_017013521.1 XP_016869010.1 6, 7 701
          XM_017013522.1 XP_016869011.1 6, 7 775
          XM_017013523.1 XP_016869012.1 6, 7 701
          XM_017013524.1 XP_016869013.1 6, 7 603
          XM_017013525.1 XP_016869014.1 6, 7 603
          XM_017013526.1 XP_016869015.1 6, 7 603
          XM_017013527.1 XP_016869016.1 5, 6 700
          XM_017013528.1 XP_016869017.1 5, 6 508
          GenBank mRNA AF151848.1 - 6, 7 707
          AK000672.1 - 6, 7 703
          AK297896.1 - 5, 6 606
          AK303122.1 - 5, 6 557
          AL561197.3 - 6, 7 587
          BC008509.1 - 4, 5 359
          BC009671.1 - 6, 7 687 3896168

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001031663, XM_006237933, BC079473, BC098713.
          This siRNA also targets the following transcripts in Mouse: NM_025476, BC025158, AK009059, AK002442, AK029446, and others.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zh286:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0