Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RMDN1 SIRNA Assays ›
Gene Symbol:
RMDN1
Gene Name:
regulator of microtubule dynamics 1
Gene Aliases:
CGI-90, FAM82B, RMD-1, RMD1
Chromosome Location:
Chr.8: 86467116 - 86514348 on Build GRCh38
UniGene ID:
Hs.145386
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126876
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RMDN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286707.1
NP_001273636.1
8
900
NM_001286719.1
NP_001273648.1
8
939
NM_001317807.1
NP_001304736.1
9
808
NM_016033.2
NP_057117.2
9
1029
XM_005250944.3
XP_005251001.1
9
935
XM_005250947.3
XP_005251004.1
8
845
XM_005250948.3
XP_005251005.1
9
837
XM_006716573.3
XP_006716636.1
9
1024
XM_011517090.2
XP_011515392.1
9
1009
XM_011517091.2
XP_011515393.1
9
970
XM_011517092.2
XP_011515394.1
8
919
XM_011517093.2
XP_011515395.1
9
1009
XM_011517094.2
XP_011515396.1
9
1009
XM_011517095.2
XP_011515397.1
9
1009
XM_011517096.2
XP_011515398.1
9
970
XM_011517097.2
XP_011515399.1
8
919
XM_011517098.2
XP_011515400.1
8
880
XM_017013518.1
XP_016869007.1
9
935
XM_017013519.1
XP_016869008.1
9
970
XM_017013520.1
XP_016869009.1
9
935
XM_017013521.1
XP_016869010.1
9
896
XM_017013524.1
XP_016869013.1
9
837
XM_017013525.1
XP_016869014.1
9
837
XM_017013526.1
XP_016869015.1
9
798
XM_017013527.1
XP_016869016.1
8
934
XM_017013528.1
XP_016869017.1
8
742
GenBank mRNA
AF151848.1
-
9
941
AK000672.1
-
9
937
AK297896.1
-
8
801
AK303122.1
-
8
791
AL561197.3
-
9
782
BC008509.1
-
7
593
BC009671.1
-
9
921
3896168
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9fbpc:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0