Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UIMC1 SIRNA Assays ›
Gene Symbol:
UIMC1
Gene Name:
ubiquitin interaction motif containing 1
Gene Aliases:
RAP80, X2HRIP110
Chromosome Location:
Chr.5: 176905005 - 177022640 on Build GRCh38
UniGene ID:
Hs.232721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126905
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UIMC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199297.2
NP_001186226.1
9
1675
NM_001199298.1
NP_001186227.1
8
1450
NM_001317961.1
NP_001304890.1
8
1063
NM_016290.4
NP_057374.3
8
1464
XM_005265930.2
XP_005265987.1
8
1438
XM_005265932.2
XP_005265989.1
6
574
XM_005265933.2
XP_005265990.1
6
686
XM_005265934.2
XP_005265991.1
8
893
XM_005265935.2
XP_005265992.1
8
902
XM_005265936.2
XP_005265993.1
7
692
XM_006714871.2
XP_006714934.1
8
1413
XM_006714874.2
XP_006714937.1
8
877
XM_011534568.2
XP_011532870.1
8
1429
XM_011534570.1
XP_011532872.1
8
1429
XM_017009574.1
XP_016865063.1
7
1329
XM_017009575.1
XP_016865064.1
9
1141
XM_017009576.1
XP_016865065.1
6
587
XM_017009577.1
XP_016865066.1
6
639
XM_017009578.1
XP_016865067.1
7
701
XM_017009579.1
XP_016865068.1
7
601
GenBank mRNA
AF113538.1
-
4
254
AF349313.1
-
8
1406
AK023044.1
-
9
1452
BC006078.1
-
8
1371
3659756
BC032561.1
-
8
854
5578134
BX537376.1
-
8
1464
EF077642.1
-
4
466
EF077644.1
-
3
204
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
double-strand break repair via nonhomologous end joining
transcription, DNA-templated
response to ionizing radiation
G2 DNA damage checkpoint
positive regulation of DNA repair
negative regulation of transcription, DNA-templated
histone H2A K63-linked deubiquitination
protein binding
histone binding
K63-linked polyubiquitin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0