Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERAP1 SIRNA Assays ›
Gene Symbol:
ERAP1
Gene Name:
endoplasmic reticulum aminopeptidase 1
Gene Aliases:
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
Chromosome Location:
Chr.5: 96759245 - 96935983 on Build GRCh38
UniGene ID:
Hs.666524
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126918
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040458.1
NP_001035548.1
3
914
NM_001198541.1
NP_001185470.1
3
672
NM_016442.3
NP_057526.3
3
914
XM_005272015.4
XP_005272072.1
3
865
XM_005272016.4
XP_005272073.1
3
825
XM_011543480.2
XP_011541782.1
7
3134
XM_011543481.2
XP_011541783.1
7
3131
XM_011543484.2
XP_011541786.1
7
3130
XM_011543485.2
XP_011541787.1
6
2950
XM_011543486.2
XP_011541788.1
7
3134
XM_017009580.1
XP_016865069.1
8
3190
XM_017009581.1
XP_016865070.1
6
2976
XM_017009582.1
XP_016865071.1
3
649
XM_017009583.1
XP_016865072.1
3
860
GenBank mRNA
AB011097.2
-
2
588
AF106037.1
-
3
629
AF183569.1
-
3
634
AF222340.1
-
3
654
AK291636.1
-
3
664
AK310535.1
-
2
89
AY028806.1
-
3
664
AY028807.1
-
3
664
AY358590.1
-
3
652
BC030775.1
-
3
666
4824919
DC398730.1
-
2
132
JX962699.1
-
2
567
JX962700.1
-
2
567
JX962701.1
-
2
567
JX962702.1
-
2
567
KM357870.1
-
2
567
KM357871.1
-
2
567
KM357872.1
-
2
567
KM357873.1
-
2
567
KM357874.1
-
2
567
KM357875.1
-
2
567
KM357876.1
-
2
567
KM357877.1
-
2
567
KM357878.1
-
2
567
KM357879.1
-
2
567
KM357880.1
-
2
567
KM357881.1
-
2
567
KM357882.1
-
2
567
KM357883.1
-
2
567
KM357884.1
-
2
567
KM357885.1
-
2
567
KM357886.1
-
2
567
KM357887.1
-
2
567
KM357888.1
-
2
567
KM357889.1
-
2
567
KM357890.1
-
2
567
KM357891.1
-
2
567
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
membrane protein ectodomain proteolysis
regulation of blood pressure
response to bacterium
antigen processing and presentation of endogenous peptide antigen via MHC class I
peptide catabolic process
regulation of innate immune response
fat cell differentiation
positive regulation of angiogenesis
endopeptidase activity
aminopeptidase activity
interleukin-6 receptor binding
interleukin-1, Type II receptor binding
tumor necrosis factor receptor binding
protein binding
metalloexopeptidase activity
zinc ion binding
peptide binding
metalloaminopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0