Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 10 | 2086 | ||
NM_001042453.1 | NP_001035918.1 | 11 | 2332 | |||
NM_016542.3 | NP_057626.2 | 12 | 2563 | |||
XM_011531349.1 | XP_011529651.1 | 13 | 2536 | |||
XM_011531350.1 | XP_011529652.1 | 12 | 2609 | |||
GenBank mRNA | AB040057.2 | - | 12 | 2483 | ||
AK000556.1 | - | 8 | 1896 | |||
AK027589.1 | - | 7 | 1780 | |||
AK074837.1 | - | 7 | 1816 | |||
AK075107.1 | - | 11 | 2244 | |||
AK075150.1 | - | 11 | 2406 | |||
BC013856.2 | - | 1 | 1048 |