Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIGG SIRNA Assays ›
Gene Symbol:
PIGG
Gene Name:
phosphatidylinositol glycan anchor biosynthesis class G
Gene Aliases:
GPI7, LAS21, MRT53, PRO4405, RLGS1930
Chromosome Location:
Chr.4: 499200 - 539921 on Build GRCh38
UniGene ID:
Hs.7099
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126998
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIGG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127178.2
NP_001120650.1
2
399
NM_001289051.1
NP_001275980.1
2
208
NM_001289052.1
NP_001275981.1
2
399
NM_001289053.1
NP_001275982.1
2
957
NM_001289055.1
NP_001275984.1
2
399
NM_001289057.1
NP_001275986.1
2
635
NM_017733.4
NP_060203.3
2
399
NR_110293.1
2
399
XM_005272284.2
XP_005272341.1
2
633
XM_011513490.2
XP_011511792.1
2
362
XM_011513491.1
XP_011511793.1
2
359
XM_011513493.1
XP_011511795.1
2
399
XM_011513494.2
XP_011511796.1
2
399
XM_017008321.1
XP_016863810.1
2
633
XM_017008322.1
XP_016863811.1
2
633
XM_017008323.1
XP_016863812.1
2
633
XM_017008324.1
XP_016863813.1
2
633
XM_017008325.1
XP_016863814.1
2
346
XM_017008326.1
XP_016863815.1
2
399
GenBank mRNA
AB162713.1
-
2
369
AK074715.1
-
2
171
AK074815.1
-
2
378
AK097244.1
-
2
596
AK296507.1
-
2
390
AK309107.1
-
2
359
AY358538.1
-
2
341
BC050262.1
-
2
869
BC110878.1
-
2
341
5750347
BC144576.1
-
2
356
DA548833.1
-
2
399
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
preassembly of GPI anchor in ER membrane
phosphotransferase activity, for other substituted phosphate groups
CP2 mannose-ethanolamine phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0