Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SORBS2 SIRNA Assays ›
          Gene Symbol:
          SORBS2
          Gene Name:
          sorbin and SH3 domain containing 2
          Gene Aliases:
          ARGBP2, PRO0618
          Chromosome Location:
          Chr.4: 185585444 - 185956716 on Build GRCh38
          UniGene ID:
          Hs.481342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 127268

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SORBS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145671.2 NP_001139143.1 8, 9 935
          NM_001145672.1 NP_001139144.1 7, 8 732
          NM_001145673.1 NP_001139145.1 8, 9 1459
          NM_001145675.1 NP_001139147.1 2, 3 426
          NM_001270771.1 NP_001257700.1 10, 11 1416
          NM_003603.6 NP_003594.3 8, 9 1249
          NM_021069.4 NP_066547.1 7, 8 1196
          XM_005263307.1 XP_005263364.1 10, 11 2053
          XM_005263308.1 XP_005263365.1 9, 10 1411
          XM_005263310.1 XP_005263367.1 9, 10 1342
          XM_005263311.1 XP_005263368.1 9, 10 1411
          XM_005263312.1 XP_005263369.1 8, 9 1249
          XM_006714388.1 XP_006714451.1 9, 10 1342
          XM_006714390.1 XP_006714453.1 8, 9 1318
          XM_017008738.1 XP_016864227.1 11, 12 2527
          XM_017008739.1 XP_016864228.1 12, 13 2830
          XM_017008740.1 XP_016864229.1 11, 12 2523
          XM_017008741.1 XP_016864230.1 11, 12 2527
          XM_017008742.1 XP_016864231.1 10, 11 2477
          XM_017008743.1 XP_016864232.1 11, 12 2458
          XM_017008744.1 XP_016864233.1 11, 12 2737
          XM_017008745.1 XP_016864234.1 10, 11 2677
          XM_017008746.1 XP_016864235.1 11, 12 2527
          XM_017008747.1 XP_016864236.1 11, 12 2668
          XM_017008748.1 XP_016864237.1 11, 12 2527
          XM_017008749.1 XP_016864238.1 11, 12 2089
          XM_017008750.1 XP_016864239.1 11, 12 2527
          XM_017008751.1 XP_016864240.1 11, 12 2527
          XM_017008752.1 XP_016864241.1 11, 12 2527
          XM_017008753.1 XP_016864242.1 11, 12 2527
          XM_017008755.1 XP_016864244.1 8, 9 1492
          XM_017008756.1 XP_016864245.1 7, 8 1195
          XM_017008758.1 XP_016864247.1 8, 9 1361
          XM_017008759.1 XP_016864248.1 8, 9 2088
          XM_017008760.1 XP_016864249.1 8, 9 1318
          XM_017008761.1 XP_016864250.1 11, 12 2527
          XM_017008762.1 XP_016864251.1 9, 10 1411
          XM_017008763.1 XP_016864252.1 8, 9 1361
          XM_017008764.1 XP_016864253.1 9, 10 1342
          XM_017008765.1 XP_016864254.1 10, 11 1645
          XM_017008766.1 XP_016864255.1 8, 9 1318
          XM_017008767.1 XP_016864256.1 9, 10 1621
          XM_017008768.1 XP_016864257.1 8, 9 1539
          XM_017008771.1 XP_016864260.1 7, 8 1199
          XM_017008772.1 XP_016864261.1 8, 9 1249
          XM_017008773.1 XP_016864262.1 9, 10 1552
          XM_017008774.1 XP_016864263.1 2, 3 826
          GenBank mRNA AB018320.1 - 7, 8 848
          AF049884.1 - 8, 9 1213
          AF049885.1 - 7, 8 578
          AK056628.1 - 10, 11 1034
          AK225812.1 - 8, 9 1459
          AK293400.1 - 7, 8 732
          AK295742.1 - 8, 9 767
          AK304653.1 - 2, 3 162
          BC011883.1 - 8, 9 902 3677070
          DC421607.1 - 2, 3 425

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          cell adhesion
          biological_process
          cell migration
          cell growth involved in cardiac muscle cell development
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal adaptor activity
          structural constituent of muscle
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-t6npl:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0