Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SORBS2 SIRNA Assays ›
          Gene Symbol:
          SORBS2
          Gene Name:
          sorbin and SH3 domain containing 2
          Gene Aliases:
          ARGBP2, PRO0618
          Chromosome Location:
          Chr.4: 185585444 - 185956716 on Build GRCh38
          UniGene ID:
          Hs.481342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 127270

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SORBS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145670.1 NP_001139142.1 18 2495
          NM_001145671.2 NP_001139143.1 20 2284
          NM_001145672.1 NP_001139144.1 18 1922
          NM_001145673.1 NP_001139145.1 20 2808
          NM_001145674.1 NP_001139146.1 12 3095
          NM_001145675.1 NP_001139147.1 12 1559
          NM_001270771.1 NP_001257700.1 21 4130
          NM_003603.6 NP_003594.3 20 2523
          NM_021069.4 NP_066547.1 18 3910
          XM_005263307.1 XP_005263364.1 22 3402
          XM_005263308.1 XP_005263365.1 21 2760
          XM_005263310.1 XP_005263367.1 21 2616
          XM_005263311.1 XP_005263368.1 20 2601
          XM_005263312.1 XP_005263369.1 20 2598
          XM_005263313.1 XP_005263370.1 18 2499
          XM_006714388.1 XP_006714451.1 21 2691
          XM_006714390.1 XP_006714453.1 19 2508
          XM_017008741.1 XP_016864230.1 24 3960
          XM_017008748.1 XP_016864237.1 21 3660
          XM_017008750.1 XP_016864239.1 24 3960
          XM_017008751.1 XP_016864240.1 23 3876
          XM_017008752.1 XP_016864241.1 23 3801
          XM_017008753.1 XP_016864242.1 21 3660
          XM_017008754.1 XP_016864243.1 20 3615
          XM_017008755.1 XP_016864244.1 18 2625
          XM_017008756.1 XP_016864245.1 17 2328
          XM_017008757.1 XP_016864246.1 13 1730
          XM_017008758.1 XP_016864247.1 21 2794
          XM_017008759.1 XP_016864248.1 18 3221
          XM_017008760.1 XP_016864249.1 20 2667
          XM_017008762.1 XP_016864251.1 19 2544
          XM_017008763.1 XP_016864252.1 18 2494
          XM_017008764.1 XP_016864253.1 19 2475
          XM_017008765.1 XP_016864254.1 20 2778
          XM_017008766.1 XP_016864255.1 18 2451
          XM_017008767.1 XP_016864256.1 19 2754
          XM_017008768.1 XP_016864257.1 18 2672
          XM_017008769.1 XP_016864258.1 18 2430
          XM_017008770.1 XP_016864259.1 17 2406
          XM_017008771.1 XP_016864260.1 19 2548
          XM_017008772.1 XP_016864261.1 18 2382
          XM_017008773.1 XP_016864262.1 19 2685
          XM_017008774.1 XP_016864263.1 14 2175
          XM_017008775.1 XP_016864264.1 23 3915
          GenBank mRNA AB018320.1 - 18 3562
          AF049884.1 - 20 2487
          AF049885.1 - 19 1927
          AK056628.1 - 21 3748
          AK056758.1 - 13 1730
          AK225327.1 - 18 2461
          AK225812.1 - 20 2808
          AK293400.1 - 18 1922
          AK296461.1 - 12 2872
          AK304653.1 - 12 1295
          BC011883.1 - 20 2251 3677070

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          cell adhesion
          biological_process
          cell migration
          cell growth involved in cardiac muscle cell development
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal adaptor activity
          structural constituent of muscle
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zq2rp:80/100.66.77.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0