Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 4 | 630 | ||
NR_033902.1 | 4 | 448 | ||||
XM_011544603.1 | XP_011542905.1 | 4 | 448 | |||
XM_011544604.1 | XP_011542906.1 | 4 | 448 | |||
XM_017013717.1 | XP_016869206.1 | 4 | 630 | |||
GenBank mRNA | AK095712.1 | - | 4 | 304 | ||
AK314535.1 | - | 4 | 323 | |||
AL834162.1 | - | 4 | 617 | |||
AL834351.1 | - | 4 | 618 | |||
AL834352.1 | - | 4 | 618 | |||
BC065495.1 | - | 4 | 306 | 5496068 | ||
BI561261.1 | - | 4 | 345 | |||
BX640952.1 | - | 4 | 317 | |||
DA721640.1 | - | 4 | 448 |