Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287497.1 | NP_001274426.1 | 3 | 1251 | ||
NM_022484.5 | NP_071929.3 | 2 | 1160 | |||
XM_017012523.1 | XP_016868012.1 | 2 | 1768 | |||
XM_017012524.1 | XP_016868013.1 | 2 | 1450 | |||
XM_017012525.1 | XP_016868014.1 | 2 | 1450 | |||
GenBank mRNA | AK025482.1 | - | 3 | 1221 | ||
AK314460.1 | - | 2 | 1127 | |||
AL136626.2 | - | 2 | 1132 | |||
BC030804.1 | - | 1 | 133 | |||
CN256103.1 | - | 1 | 254 |