Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024091.3 | NP_076996.2 | 2 | 571 | ||
XM_006714498.1 | XP_006714561.1 | 2 | 482 | |||
GenBank mRNA | AK026927.1 | - | 2 | 536 | ||
BC001295.1 | - | 1 | 63 | |||
BC113563.1 | - | 1 | 486 | 8322615 | ||
BC143517.1 | - | 1 | 486 | |||
BQ921348.1 | - | 2 | 528 |