Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178055.1 | NP_001171526.1 | 13 | 1161 | ||
NM_001178056.1 | NP_001171527.1 | 12 | 1137 | |||
NM_024615.3 | NP_078891.2 | 12 | 1137 | |||
XM_005248596.4 | XP_005248653.1 | 13 | 1517 | |||
XM_011543631.2 | XP_011541933.1 | 13 | 1050 | |||
XM_011543632.2 | XP_011541934.2 | 13 | 1097 | |||
XM_011543633.2 | XP_011541935.2 | 13 | 1097 | |||
XM_011543634.2 | XP_011541936.1 | 14 | 1646 | |||
XM_011543635.2 | XP_011541937.1 | 14 | 1167 | |||
XM_011543636.2 | XP_011541938.1 | 13 | 1031 | |||
XM_011543640.2 | XP_011541942.1 | 10 | 921 | |||
XM_011543641.2 | XP_011541943.1 | 11 | 898 | |||
XM_011543642.2 | XP_011541944.1 | 9 | 885 | |||
XM_011543643.2 | XP_011541945.1 | 10 | 1326 | |||
XM_017009850.1 | XP_016865339.1 | 12 | 1597 | |||
XM_017009851.1 | XP_016865340.1 | 11 | 1037 | |||
GenBank mRNA | AK289369.1 | - | 12 | 1120 | ||
AK298680.1 | - | 11 | 1000 | |||
AK314466.1 | - | 13 | 1161 | |||
AL834477.1 | - | 12 | 1096 | |||
BC037386.1 | - | 10 | 995 | 4830820 | ||
BC075801.1 | - | 12 | 1096 | 5226828 |