Search Thermo Fisher Scientific
Gene Symbol: CEP135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025009.4 | NP_079285.2 | 2, 3 | 418 | ||
XM_005265788.3 | XP_005265845.1 | 2, 3 | 389 | |||
XM_006714055.2 | XP_006714118.1 | 2, 3 | 418 | |||
GenBank mRNA | AK023683.1 | - | 2, 3 | 355 | ||
BC136535.1 | - | 2, 3 | 245 | 9020525 | ||
BC136536.1 | - | 2, 3 | 245 | 9020526 | ||
DA944890.1 | - | 2, 3 | 418 |