Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030782.4 | NP_110409.2 | 17 | 1789 | ||
XM_011514144.2 | XP_011512446.1 | 17 | 1753 | |||
GenBank mRNA | AB045223.1 | - | 17 | 1546 | ||
AK027306.1 | - | 17 | 1603 | |||
AK057095.1 | - | 1 | 1025 | |||
AK126225.1 | - | 1 | 2907 | |||
AK128462.1 | - | 8 | 2982 | |||
AL137440.1 | - | 1 | 124 | |||
AL832953.1 | - | 7 | 391 | |||
BC016399.1 | - | 14 | 1067 | |||
BC025305.1 | - | 17 | 1607 | 3051368 | ||
BC040705.1 | - | 1 | 915 |