Search
Search

CYSTM1| Gene Symbol: CYSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032412.3 | NP_115788.1 | 3 | 881 | ||
| GenBank mRNA | AJ245877.1 | - | 3 | 771 | ||
| AK225992.1 | - | 3 | 761 | |||
| BC013643.1 | - | 3 | 791 | 4152168 | ||
| BC023982.1 | - | 3 | 798 | 4046171 | ||