Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032847.2 | NP_116236.1 | 4 | 431 | ||
GenBank mRNA | AK027731.1 | - | 4 | 410 | ||
AK222660.1 | - | 4 | 379 | |||
BC067796.1 | - | 4 | 405 | 5260773 | ||
DR156993.1 | - | 4 | 431 |
Gene Symbol: ZHX1-C8orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204180.1 | NP_001191109.1 | 5 | 647 | ||
GenBank mRNA | BC012379.2 | - | 4 | 377 | 3857253 | |
BP340117.1 | - | 5 | 444 |