Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033035.4 | NP_149024.1 | 4 | 891 | ||
NM_138551.4 | NP_612561.2 | 2 | 673 | |||
NR_045089.1 | 5 | 2096 | ||||
XM_011543698.1 | XP_011542000.1 | 3 | 495 | |||
GenBank mRNA | AL833421.1 | - | 2 | 653 | ||
BC016720.1 | - | 2 | 606 | |||
BC022894.1 | - | 2 | 645 | |||
BC040592.1 | - | 5 | 2097 | 5269764 |