Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133468.4 | NP_597725.1 | 8 | 1012 | ||
XM_005249633.2 | XP_005249690.1 | 7 | 752 | |||
GenBank mRNA | AB075845.1 | - | 4 | 279 | ||
AK289963.1 | - | 7 | 752 | |||
AY324650.1 | - | 8 | 1011 | |||
BC060868.1 | - | 7 | 689 | 30345290 |