Search
Search

CMBL| Gene Symbol: CMBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138809.3 | NP_620164.1 | 6 | 1214 | ||
| XM_017009034.1 | XP_016864523.1 | 6 | 978 | |||
| XM_017009035.1 | XP_016864524.1 | 6 | 934 | |||
| GenBank mRNA | AJ278125.1 | - | 6 | 1007 | ||
| AK074197.1 | - | 6 | 1005 | |||
| BC001573.1 | - | 6 | 920 | 3454199 | ||
| BX647141.1 | - | 4 | 486 | |||