Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139166.4 | NP_631905.1 | 2 | 1181 | ||
GenBank mRNA | AF503617.1 | - | 2 | 1127 | ||
AK092694.1 | - | 2 | 1183 | |||
AL832152.1 | - | 2 | 1182 | |||
AL833422.1 | - | 2 | 1180 | |||
BC105103.1 | - | 2 | 1169 | 8144106 | ||
BC105105.1 | - | 2 | 1169 | 8144108 |