Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145030.2 | NP_659467.1 | 4 | 722 | ||
XM_011515914.1 | XP_011514216.1 | 4 | 469 | |||
GenBank mRNA | BC026269.1 | - | 4 | 722 | 4773899 |