Search
Search

MARCH3| Gene Symbol: MARCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178450.4 | NP_848545.1 | 5 | 1902 | ||
| XM_011543128.2 | XP_011541430.1 | 4 | 1639 | |||
| XM_011543129.1 | XP_011541431.1 | 6 | 1767 | |||
| XM_011543130.1 | XP_011541432.1 | 6 | 1672 | |||
| XM_011543131.2 | XP_011541433.1 | 6 | 1901 | |||
| XM_017009014.1 | XP_016864503.1 | 4 | 2090 | |||
| GenBank mRNA | AF055007.1 | - | 5 | 1562 | ||
| BC047569.1 | - | 5 | 1653 | 4819831 | ||