Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182491.2 | NP_872297.2 | 5 | 797 | ||
GenBank mRNA | AK290170.1 | - | 5 | 756 | ||
BC016963.1 | - | 5 | 749 | |||
BC029558.1 | - | 5 | 740 | 5263788 |