Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR1 SIRNA Assays ›
          Gene Symbol:
          TLR1
          Gene Name:
          toll like receptor 1
          Gene Aliases:
          CD281, TIL, TIL. LPRS5, rsc786
          Chromosome Location:
          Chr.4: 38787555 - 38805800 on Build GRCh38
          UniGene ID:
          Hs.621817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 12857

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003263.3 NP_003254.2 4 780
          XM_005262662.4 XP_005262719.1 5 1465
          XM_011513742.2 XP_011512044.1 4 1388
          XM_011513743.2 XP_011512045.1 4 1410
          XM_011513744.2 XP_011512046.1 3 1333
          XM_011513745.2 XP_011512047.1 3 1296
          XM_017008571.1 XP_016864060.1 4 1431
          XM_017008572.1 XP_016864061.1 4 1373
          GenBank mRNA AB445617.1 - 1 506
          AK292982.1 - 4 791
          AL050262.1 - 4 780
          BC089403.1 - 3 736 3063206
          BC109093.1 - 2 601 40034681
          BC109094.1 - 2 601 40034683
          CR533562.1 - 1 506
          D13637.1 - 1 569
          DQ012254.1 - 1 506
          DQ012255.1 - 1 506
          DQ012256.1 - 1 506
          DQ012257.1 - 1 506
          DQ012258.1 - 1 506
          DQ012259.1 - 1 506
          DQ012260.1 - 1 506
          DQ012261.1 - 1 506
          DQ012262.1 - 1 506
          DQ012263.1 - 1 506
          DQ012264.1 - 1 506
          U88540.1 - 1 506

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          toll-like receptor signaling pathway
          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          immune response
          signal transduction
          toll-like receptor 1 signaling pathway
          toll-like receptor TLR1:TLR2 signaling pathway
          macrophage activation
          detection of triacyl bacterial lipopeptide
          positive regulation of tumor necrosis factor biosynthetic process
          defense response to bacterium
          innate immune response
          positive regulation of interleukin-6 biosynthetic process
          regulation of cytokine secretion
          cellular response to triacyl bacterial lipopeptide
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          Toll-like receptor 2 binding
          protein heterodimerization activity
          lipopeptide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0