Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF216 SIRNA Assays ›
          Gene Symbol:
          RNF216 RNF216P1
          Gene Name:
          ring finger protein 216
          ring finger protein 216 pseudogene 1
          Gene Aliases:
          CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN
          RNF216L, hCG2040556
          Chromosome Location:
          Chr.7: 5620041 - 5781730 on Build GRCh38
          Chromosome Location:
          Chr.7: 4973985 - 4998169 on Build GRCh38
          UniGene ID:
          Hs.487458
          Hs.520636
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 128671

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RNF216
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_207111.3 NP_996994.1 7 1623
          NM_207116.2 NP_996999.1 7 1452
          XM_005249785.2 XP_005249842.1 9 1943
          XM_011515436.1 XP_011513738.1 5 1362
          XM_017012363.1 XP_016867852.1 7 1307
          XM_017012364.1 XP_016867853.1 7 1596
          XM_017012365.1 XP_016867854.1 6 1265
          GenBank mRNA AF513717.1 - 7 1288
          AF513718.1 - 7 1459
          AK092767.1 - 7 1538
          AK123635.1 - 4 777
          AK292396.1 - 7 1632
          AK297416.1 - 8 807
          AK314332.1 - 4 404
          AY062174.1 - 4 469
          AY177396.1 - 4 527
          AY177397.1 - 6 1537
          AY177398.1 - 6 1541
          BC000787.2 - 1 37
          BC004947.1 - 7 1342
          BC063825.1 - 7 1362 6504476
          BX537406.1 - 7 1384
          Gene Symbol: RNF216P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015449.1 4 557
          NR_023384.1 4 557
          NR_023385.1 4 557
          GenBank mRNA AK303831.1 - 4 408
          AK308526.1 - 4 492
          AK311517.1 - 4 490
          BC026736.1 - 4 436
          BC068459.1 - 4 526
          BX538059.1 - 4 484
          DC357738.1 - 4 557

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          viral process
          negative regulation of type I interferon production
          regulation of interferon-beta production
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of defense response to virus by host
          protein K48-linked ubiquitination
          protein linear polyubiquitination
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          ubiquitin binding
          metal ion binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0