Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS2 SIRNA Assays ›
Gene Symbol:
CADPS2
Gene Name:
calcium dependent secretion activator 2
Gene Aliases:
CAPS2
Chromosome Location:
Chr.7: 122318424 - 122886759 on Build GRCh38
UniGene ID:
Hs.649459
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128730
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009571.3
NP_001009571.2
28
4331
NM_001167940.1
NP_001161412.1
30
4466
NM_017954.10
NP_060424.9
30
4454
XM_005250695.4
XP_005250752.1
32
4219
XM_005250696.4
XP_005250753.1
31
4210
XM_005250697.4
XP_005250754.1
31
4210
XM_005250698.4
XP_005250755.1
31
4176
XM_005250699.4
XP_005250756.1
31
4198
XM_005250700.4
XP_005250757.1
30
4195
XM_005250701.4
XP_005250758.1
30
4189
XM_005250702.4
XP_005250759.1
29
4174
XM_005250703.4
XP_005250760.1
31
4099
XM_005250704.4
XP_005250761.1
30
4090
XM_005250705.4
XP_005250762.1
29
4075
XM_005250706.4
XP_005250763.1
29
4075
XM_005250707.4
XP_005250764.1
29
4069
XM_005250708.3
XP_005250765.1
27
4045
XM_011516695.2
XP_011514997.1
30
4056
XM_011516696.2
XP_011514998.1
29
4063
XM_011516697.2
XP_011514999.1
28
4054
XM_017012794.1
XP_016868283.1
29
4180
XM_017012795.1
XP_016868284.1
28
4066
XM_017012796.1
XP_016868285.1
33
3834
XM_017012797.1
XP_016868286.1
32
3885
XM_017012798.1
XP_016868287.1
31
4349
XM_017012799.1
XP_016868288.1
31
4136
XM_017012800.1
XP_016868289.1
29
3227
GenBank mRNA
AB046811.1
-
26
3200
AF401638.1
-
28
4000
AK000768.1
-
13
1509
AK054853.1
-
15
1613
AK126518.1
-
29
3933
AY264289.1
-
30
4058
BC136601.1
-
28
4030
9020591
BC144278.1
-
28
4030
9052798
BC144279.1
-
30
4165
9052799
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to starvation
protein transport
synaptic vesicle priming
positive regulation of exocytosis
dense core granule exocytosis
lipid binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0