Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 4 | 1017 | ||
NM_181268.2 | NP_851785.2 | 4 | 1001 | |||
NM_181269.2 | NP_851786.1 | 4 | 722 | |||
NM_181270.2 | NP_851787.1 | 4 | 706 | |||
NM_181271.2 | NP_851788.1 | 3 | 581 | |||
NM_181272.2 | NP_851789.1 | 3 | 563 | |||
NM_181283.2 | NP_851800.1 | 3 | 547 | |||
NM_181296.2 | NP_851813.1 | 2 | 448 | |||
GenBank mRNA | AF278576.1 | - | 4 | 621 | ||
AF278577.1 | - | 4 | 637 | |||
AY174118.1 | - | 4 | 726 | |||
AY174119.1 | - | 4 | 710 | |||
AY174120.1 | - | 3 | 585 | |||
AY174121.1 | - | 3 | 567 | |||
AY174122.1 | - | 3 | 551 | |||
AY174123.1 | - | 3 | 722 | |||
AY174124.1 | - | 3 | 706 | |||
AY174125.1 | - | 5 | 860 | |||
AY174126.1 | - | 5 | 844 | |||
AY174127.1 | - | 2 | 414 | |||
AY174128.1 | - | 4 | 690 | |||
AY174129.1 | - | 5 | 713 | |||
AY174130.1 | - | 5 | 885 | |||
AY174131.1 | - | 3 | 542 | |||
AY174132.1 | - | 3 | 526 | |||
AY174133.1 | - | 2 | 452 | |||
AY174134.1 | - | 4 | 772 | |||
AY174135.1 | - | 5 | 950 | |||
AY174136.1 | - | 5 | 934 | |||
AY174137.1 | - | 4 | 752 | |||
AY174138.1 | - | 4 | 736 | |||
BC057852.1 | - | 4 | 1017 | 5273108 | ||
DQ309766.1 | - | 5 | 835 | |||
DQ355985.1 | - | 3 | 573 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 3 | 590 | ||
NM_001204098.1 | NP_001191027.1 | 4 | 670 | |||
NM_001204099.1 | NP_001191028.1 | 4 | 749 |