Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TJAP1 SIRNA Assays ›
Gene Symbol:
TJAP1
Gene Name:
tight junction associated protein 1
Gene Aliases:
PILT, TJP4
Chromosome Location:
Chr.6: 43477523 - 43506557 on Build GRCh38
UniGene ID:
Hs.520145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128980
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TJAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146016.1
NP_001139488.1
11
1774
NM_001146017.1
NP_001139489.1
10
1739
NM_001146018.1
NP_001139490.1
11
1801
NM_001146019.1
NP_001139491.1
10
1744
NM_001146020.1
NP_001139492.1
9
1595
NM_080604.2
NP_542171.2
10
1796
NR_027394.1
10
1661
XM_006715249.1
XP_006715312.1
11
1752
XM_006715250.3
XP_006715313.1
12
1771
XM_006715251.2
XP_006715314.1
11
1779
XM_006715252.2
XP_006715315.1
12
1839
XM_006715254.3
XP_006715317.1
11
1677
XM_006715256.1
XP_006715319.1
11
1674
XM_006715257.2
XP_006715320.1
11
1728
XM_006715259.3
XP_006715322.1
10
1578
XM_006715260.1
XP_006715323.1
11
1695
XM_006715261.1
XP_006715324.1
10
1599
XM_006715262.2
XP_006715325.1
11
1924
XM_006715263.2
XP_006715326.1
10
1634
XM_006715265.1
XP_006715328.1
10
1644
XM_006715266.1
XP_006715329.1
9
1552
XM_006715267.2
XP_006715330.1
11
1763
XM_006715268.2
XP_006715331.1
11
1760
XM_006715269.2
XP_006715332.1
11
1815
XM_006715270.1
XP_006715333.1
10
1733
XM_011514995.1
XP_011513297.1
12
1814
XM_011514996.1
XP_011513298.1
11
1734
XM_017011488.1
XP_016866977.1
10
1652
XM_017011489.1
XP_016866978.1
11
1949
XM_017011490.1
XP_016866979.1
10
1722
XM_017011491.1
XP_016866980.1
11
1758
XM_017011492.1
XP_016866981.1
12
1887
XM_017011493.1
XP_016866982.1
12
1885
GenBank mRNA
AJ420477.1
-
2
895
AK024269.1
-
10
1746
AK074241.1
-
5
1190
AK094907.1
-
3
1602
AK123398.1
-
7
4304
AK127338.1
-
9
2857
AK225787.1
-
10
1697
AK309119.1
-
10
1614
AL117398.1
-
4
2228
BC046239.1
-
11
1694
4906152
BC064401.1
-
10
1675
5539930
CR749480.1
-
8
4125
Back To Top
More Information
Panther Classification:
Molecular Function -
tight junction
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0