Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139284.2 | NP_644813.1 | 9 | 2422 | ||
XM_011526595.1 | XP_011524897.1 | 10 | 1903 | |||
XM_017026428.1 | XP_016881917.1 | 9 | 1751 | |||
XM_017026429.1 | XP_016881918.1 | 10 | 1823 | |||
XM_017026430.1 | XP_016881919.1 | 8 | 1605 | |||
GenBank mRNA | AJ487519.1 | - | 9 | 2347 | ||
AJ487959.1 | - | 9 | 1855 | |||
AL832968.1 | - | 6 | 2323 | |||
BC029943.1 | - | 10 | 1887 | |||
BC136694.1 | - | 9 | 1897 | 9020684 | ||
BC136697.1 | - | 9 | 1897 | 9020687 |