Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CFAP70 SIRNA Assays ›
Gene Symbol:
CFAP70
Gene Name:
cilia and flagella associated protein 70
Gene Aliases:
TTC18
Chromosome Location:
Chr.10: 73253758 - 73361064 on Build GRCh38
UniGene ID:
Hs.591367
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
129332
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CFAP70
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_145170.3
NP_660153.3
17
2018
XM_005269489.2
XP_005269546.1
16
1888
XM_006717604.2
XP_006717667.1
18
3579
XM_006717605.2
XP_006717668.1
17
1981
XM_006717609.2
XP_006717672.1
15
1795
XM_006717610.2
XP_006717673.1
16
3267
XM_006717611.3
XP_006717674.1
16
1919
XM_011539212.2
XP_011537514.1
13
1550
XM_017015620.1
XP_016871109.1
18
3425
XM_017015621.1
XP_016871110.1
18
3422
XM_017015622.1
XP_016871111.1
18
3422
XM_017015623.1
XP_016871112.1
18
3425
XM_017015624.1
XP_016871113.1
17
3344
XM_017015625.1
XP_016871114.1
18
3425
XM_017015626.1
XP_016871115.1
18
3425
XM_017015627.1
XP_016871116.1
17
3215
XM_017015628.1
XP_016871117.1
16
1797
XM_017015629.1
XP_016871118.1
18
3425
XM_017015630.1
XP_016871119.1
17
3499
XM_017015631.1
XP_016871120.1
13
1356
XM_017015632.1
XP_016871121.1
16
3394
XM_017015633.1
XP_016871122.1
16
1864
GenBank mRNA
AK098631.1
-
2
118
AL833537.1
-
15
5476
BC032856.2
-
17
2013
5267050
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0