Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152309.2 | NP_689522.2 | 17 | 2953 | ||
XM_005269498.1 | XP_005269555.1 | 16 | 2476 | |||
XM_005269499.1 | XP_005269556.1 | 16 | 2500 | |||
XM_011539248.1 | XP_011537550.1 | 16 | 2826 | |||
XM_011539249.1 | XP_011537551.1 | 16 | 2789 | |||
GenBank mRNA | AK092883.1 | - | 16 | 2476 | ||
AK122584.1 | - | 6 | 3782 | |||
AK125635.1 | - | 17 | 2945 | |||
BX648550.1 | - | 17 | 2849 |