Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152731.2 | NP_689944.2 | 6 | 1371 | ||
XM_005248889.1 | XP_005248946.1 | 4 | 539 | |||
XM_017010406.1 | XP_016865895.1 | 6 | 922 | |||
XM_017010407.1 | XP_016865896.1 | 6 | 905 | |||
XM_017010408.1 | XP_016865897.1 | 7 | 1452 | |||
GenBank mRNA | AK054724.1 | - | 7 | 953 | ||
AK303705.1 | - | 5 | 741 | |||
AK315926.1 | - | 3 | 335 | |||
BC022988.1 | - | 5 | 766 | 4932760 |