Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM26D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256887.2 | NP_001243816.1 | 4 | 3487 | ||
NM_001256888.2 | NP_001243817.1 | 4 | 3393 | |||
NM_001256889.2 | NP_001243818.1 | 3 | 3271 | |||
NM_153036.4 | NP_694581.1 | 3 | 3326 | |||
XM_005266860.3 | XP_005266917.1 | 2 | 4261 | |||
XM_011535560.2 | XP_011533862.1 | 5 | 3581 | |||
XM_011535561.2 | XP_011533863.1 | 4 | 3430 | |||
XM_011535562.2 | XP_011533864.1 | 6 | 3762 | |||
XM_011535563.2 | XP_011533865.1 | 4 | 3375 | |||
XM_011535564.1 | XP_011533866.1 | 4 | 3336 | |||
XM_017010390.1 | XP_016865879.1 | 5 | 3509 | |||
XM_017010391.1 | XP_016865880.1 | 5 | 3464 | |||
XM_017010392.1 | XP_016865881.1 | 5 | 3526 | |||
GenBank mRNA | AK056801.1 | - | 4 | 1440 |