Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135993.1 | NP_001129465.1 | 6 | 1423 | ||
NM_153211.3 | NP_694943.2 | 6 | 1070 | |||
XM_005258198.4 | XP_005258255.1 | 8 | 1457 | |||
XM_006722392.3 | XP_006722455.1 | 6 | 1611 | |||
XM_011525814.2 | XP_011524116.1 | 6 | 1611 | |||
XM_017025550.1 | XP_016881039.1 | 8 | 1551 | |||
GenBank mRNA | AK091080.1 | - | 6 | 1070 | ||
BC032684.1 | - | 6 | 872 | |||
BC121034.2 | - | 6 | 953 | 40117554 | ||
BC121035.2 | - | 6 | 953 | 40117556 |