Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153330.4 | NP_699161.1 | 3 | 2019 | ||
XM_006713519.3 | XP_006713582.1 | 2 | 2916 | |||
GenBank mRNA | AK125946.1 | - | 2 | 2916 | ||
AK126068.1 | - | 1 | 594 | |||
BC029521.1 | - | 1 | 612 | 5296554 |