Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153487.3 | NP_705691.1 | 17 | 5093 | ||
XM_017010734.1 | XP_016866223.1 | 17 | 4055 | |||
GenBank mRNA | AF478693.1 | - | 17 | 5093 | ||
AK126965.1 | - | 3 | 2535 | |||
BX647180.1 | - | 5 | 1672 | |||
BX647253.1 | - | 5 | 1672 |