Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286367.1 | NP_001273296.1 | 7 | 1468 | ||
NM_174913.2 | NP_777573.1 | 7 | 1468 | |||
XM_005267385.1 | XP_005267442.1 | 7 | 1495 | |||
XM_011536526.1 | XP_011534828.1 | 7 | 1287 | |||
GenBank mRNA | AB095941.1 | - | 7 | 1460 | ||
AK123717.1 | - | 2 | 243 | |||
BC025332.1 | - | 7 | 1446 | 4549328 | ||
BX248018.1 | - | 7 | 1430 | |||
BX647311.1 | - | 7 | 1435 |