Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286615.1 | NP_001273544.1 | 4 | 829 | ||
NM_001286616.1 | NP_001273545.1 | 3 | 397 | |||
NM_178826.3 | NP_849148.2 | 3 | 724 | |||
XM_011537911.2 | XP_011536213.1 | 6 | 824 | |||
XM_011537912.2 | XP_011536214.1 | 6 | 824 | |||
XM_011537913.2 | XP_011536215.1 | 6 | 824 | |||
XM_011537914.2 | XP_011536216.1 | 3 | 335 | |||
XM_011537915.2 | XP_011536217.1 | 6 | 824 | |||
XM_011537916.2 | XP_011536218.1 | 4 | 488 | |||
XM_011537918.2 | XP_011536220.1 | 6 | 824 | |||
XM_017018808.1 | XP_016874297.1 | 3 | 335 | |||
XM_017018809.1 | XP_016874298.1 | 5 | 915 | |||
XM_017018810.1 | XP_016874299.1 | 3 | 577 | |||
XM_017018811.1 | XP_016874300.1 | 3 | 577 | |||
XM_017018812.1 | XP_016874301.1 | 4 | 2727 | |||
XM_017018813.1 | XP_016874302.1 | 3 | 577 | |||
XM_017018814.1 | XP_016874303.1 | 4 | 2727 | |||
XM_017018815.1 | XP_016874304.1 | 4 | 2727 | |||
XM_017018816.1 | XP_016874305.1 | 4 | 810 | |||
XM_017018817.1 | XP_016874306.1 | 4 | 2727 | |||
GenBank mRNA | AK091591.1 | - | 3 | 513 | ||
AK293810.1 | - | 4 | 467 | |||
AK302962.1 | - | 6 | 755 | |||
AK316105.1 | - | 4 | 468 | |||
BC109308.2 | - | 3 | 382 | 40035759 | ||
DA213819.1 | - | 1 | 75 |