Search
Search

UGCG| Gene Symbol: UGCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003358.2 | NP_003349.1 | 2 | 578 | ||
| XM_005252186.4 | XP_005252243.1 | 2 | 523 | |||
| XM_017015107.1 | XP_016870596.1 | 2 | 237 | |||
| GenBank mRNA | AK314847.1 | - | 2 | 183 | ||
| BC038711.1 | - | 2 | 579 | 5295561 | ||
| D50840.1 | - | 2 | 418 | |||