Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142800.1 | NP_001136272.1 | 4 | 622 | ||
NM_001142801.1 | NP_001136273.1 | 4 | 622 | |||
NM_001292009.1 | NP_001278938.1 | 4 | 622 | |||
NM_198283.1 | NP_938024.1 | 3 | 481 | |||
GenBank mRNA | AB593112.1 | - | 4 | 578 | ||
AB593114.1 | - | 4 | 587 | |||
AK310478.1 | - | 6 | 654 | |||
AY358133.1 | - | 3 | 481 | |||
BC133011.1 | - | 2 | 346 | |||
BC133013.1 | - | 2 | 343 | |||
BX537914.1 | - | 4 | 622 | |||
BX647741.1 | - | 4 | 612 | |||
FJ416331.1 | - | 4 | 630 | |||
FM209056.1 | - | 4 | 574 |