Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMS22L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198468.2 | NP_940870.2 | 5 | 619 | ||
XM_006715430.1 | XP_006715493.1 | 5 | 550 | |||
XM_006715432.3 | XP_006715495.1 | 5 | 550 | |||
XM_011535670.2 | XP_011533972.1 | 5 | 550 | |||
XM_011535671.2 | XP_011533973.1 | 5 | 646 | |||
XM_011535672.2 | XP_011533974.1 | 5 | 613 | |||
XM_011535673.2 | XP_011533975.1 | 5 | 619 | |||
XM_011535674.2 | XP_011533976.1 | 5 | 515 | |||
XM_011535675.2 | XP_011533977.1 | 5 | 550 | |||
XM_017010656.1 | XP_016866145.1 | 4 | 548 | |||
XM_017010657.1 | XP_016866146.1 | 4 | 547 | |||
GenBank mRNA | AK128060.1 | - | 5 | 619 | ||
BX538214.1 | - | 1 | 56 | |||
CR749822.1 | - | 5 | 569 |