Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282772.1 | NP_001269701.1 | 2, 3 | 415 | ||
NM_001282774.1 | NP_001269703.1 | 1, 2 | 765 | |||
NM_001282777.1 | NP_001269706.1 | 2, 3 | 857 | |||
NM_019014.5 | NP_061887.2 | 1, 2 | 765 | |||
GenBank mRNA | AK001678.1 | - | 1, 2 | 237 | ||
AK128044.1 | - | 1, 2 | 765 | |||
AK302814.1 | - | 2, 3 | 415 | |||
BC110833.1 | - | 1, 2 | 454 | 6053210 | ||
BX647360.1 | - | 2, 3 | 583 |