siRNA Details
Gene Symbol: POLR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137604.2 | NP_001131076.1 | 7 | 1591 | ||
NM_001282772.1 | NP_001269701.1 | 9 | 1409 | |||
NM_001282776.1 | NP_001269705.1 | 5 | 1311 | |||
NM_001282777.1 | NP_001269706.1 | 9 | 1851 | |||
NM_001282779.1 | NP_001269708.1 | 8 | 1688 | |||
NM_019014.5 | NP_061887.2 | 8 | 1759 | |||
GenBank mRNA | AK001678.1 | - | 8 | 1231 | ||
AK022533.1 | - | 7 | 1041 | |||
AK128044.1 | - | 8 | 1759 | |||
AK294027.1 | - | 8 | 1160 | |||
AK301178.1 | - | 5 | 783 | |||
AK302814.1 | - | 9 | 1409 | |||
BX647360.1 | - | 9 | 1577 | |||
BX647683.1 | - | 8 | 1138 |

More Information
Panther Classification:
Molecular Function -





