Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310156.1 | NP_001297085.1 | 6 | 782 | ||
XM_011542514.2 | XP_011540816.1 | 6 | 782 | |||
XM_011542515.2 | XP_011540817.1 | 6 | 782 | |||
XM_011542516.1 | XP_011540818.1 | 6 | 782 | |||
XM_011542517.1 | XP_011540819.1 | 6 | 782 | |||
XM_011542519.2 | XP_011540821.1 | 6 | 782 | |||
XM_011542520.2 | XP_011540822.1 | 6 | 782 | |||
XM_011542521.2 | XP_011540823.1 | 6 | 782 | |||
XM_011542522.2 | XP_011540824.1 | 6 | 782 | |||
GenBank mRNA | AF306347.1 | - | 6 | 754 | ||
BC119641.1 | - | 6 | 686 | |||
BC119642.1 | - | 6 | 686 | |||
BC127634.1 | - | 6 | 686 | 40115717 |