Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCLRE1C SIRNA Assays ›
Gene Symbol:
DCLRE1C
Gene Name:
DNA cross-link repair 1C
Gene Aliases:
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
Chromosome Location:
Chr.10: 14897853 - 14954432 on Build GRCh38
UniGene ID:
Hs.655932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130366
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCLRE1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033855.2
NP_001029027.1
10
1287
NM_001033857.2
NP_001029029.1
11
1372
NM_001033858.2
NP_001029030.1
12
1694
NM_001289076.1
NP_001276005.1
8
1146
NM_001289077.1
NP_001276006.1
11
1418
NM_001289078.1
NP_001276007.1
8
1179
NM_001289079.1
NP_001276008.1
12
1740
NM_022487.3
NP_071932.2
9
1231
NR_110297.1
12
1499
XM_006717491.3
XP_006717554.1
10
961
XM_011519616.1
XP_011517918.1
10
977
XM_011519617.1
XP_011517919.1
11
1096
XM_011519619.1
XP_011517921.1
12
1123
XM_011519620.2
XP_011517922.1
10
962
XM_011519621.1
XP_011517923.1
10
962
XM_017016556.1
XP_016872045.1
10
962
XM_017016557.1
XP_016872046.1
10
989
XM_017016558.1
XP_016872047.1
11
1035
XM_017016559.1
XP_016872048.1
8
830
XM_017016560.1
XP_016872049.1
11
1051
GenBank mRNA
AF395747.1
-
11
988
AF395748.1
-
11
1034
AF395749.1
-
9
847
AF395750.1
-
8
795
AF395751.1
-
12
1310
AF395752.1
-
12
1356
AJ296101.1
-
10
903
AK021422.1
-
9
894
AK022922.1
-
12
1293
AK290598.1
-
10
950
BC009185.2
-
8
770
3945623
BC022254.1
-
10
887
4272155
BX648858.1
-
11
1004
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
double-strand break repair via nonhomologous end joining
response to ionizing radiation
B cell differentiation
protection from non-homologous end joining at telomere
V(D)J recombination
interstrand cross-link repair
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
damaged DNA binding
endonuclease activity
protein binding
5'-3' exonuclease activity
5'-3' exodeoxyribonuclease activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0